Learn More
Abcam BC-11-38, PDE-11 inhibitor, 10MG
Supplier:  Abcam AB14615610MG
Encompass_Preferred
MW 304.43 g/mol. Potent, selective inhibitor phosphodiesterase PDE-11 inhibitor (IC₅₀ = 0.28 μM). IC₅₀s for PDE1-10 are greater than 100 μM. Significantly elevates cAMP levels and cortisol production in H295R human adenocarcinoma cells.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4910