Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BCA-1/CXCL13 Human 100ug
SDP

Supplier:  Medchemexpress LLC HYP7131100UG

Encompass_Preferred

BCA-1/CXCL13 Protein, Human is a recombinant, tag-free chemokine produced in Escherichia coli and supplied as a lyophilized material for in vitro research. CXCL13 selectively attracts B lymphocytes via the CXCR5 receptor and has validated biological activity in chemotaxis and cell proliferation assays, making it suitable for functional and signaling studies.

  • Recombinant human CXCL13 produced in Escherichia coli.
  • Tag free, lyophilized preparation for stable storage.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Approximate molecular weight ~11 kDa under reducing conditions.
  • Demonstrated chemotactic activity for human B cells; ED50 typically ~4.6 ng/mL.
  • Supplied as a 100 μg vial suitable for in vitro assays.
  • Store at -20°C; aliquot and freeze at -80°C for extended storage.

Catalog No. 50-001-96889


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.