Learn More
Medchemexpress LLC BCA-1/CXCL13 Mouse 1mg | 1mg
SDP
Supplier:  Medchemexpress LLC HYP779121MG
Encompass_Preferred
CXCL13 known as BCA-1 (B cell-attracting chemokine 1) or BLC (B-lymphocyte chemoattractant) is an efficacious attractant selective for B lymphocytes through binding to the BLR1/CXCR5 receptor[1] CXCL13 is a homeostatic chemokine and is constitutively secreted by stromal cells in B-cell areas of secondary lymphoid tissues (follicles) such as spleen lymph nodes tonsils and Peyer s patches[2] BCA-1/CXCL13 Protein Mouse (HEK293 Fc) is produced in HEK293 cells with a N-Terminal Fc-tag It consists of 88 amino acids (I22-A109)
Catalog No. 50-001-87589