Learn More
Medchemexpress LLC BCA-1/CXCL13 protein, cynomolgus (His-tag) | >95.0% | 1 MG

Supplier: Medchemexpress LLC HYP761641MG
BCA-1/CXCL13 protein (cynomolgus) is a recombinant chemokine supplied lyophilized for in vitro research applications such as cell migration and receptor-binding assays. The protein is provided with an N-terminal His-tag and comes with purity and activity specifications suitable for functional studies.
- Recombinant cynomolgus CXCL13 with N-terminal His-tag.
- Produced in E. coli expression system and supplied lyophilized.
- Purity greater than 95% as determined by reducing SDS-PAGE.
- Formulated in 10 mM Tris, 250 mM NaCl, pH 8.0; reconstitute at ≥100 μg/mL.
- Chemoattractant activity for Ramos cells with ED50 ≈0.019 μg/mL.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.