Learn More
AdipoGen BCECF acid
SDP
Supplier:  AdipoGen CDXC0002M025
Encompass_Preferred
Chemical. CAS 85138-49-4. Formula C27H20O11. MW 520.4. Synthetic. Fluorescent indicator for measurement of cytoplasmic pH in lymphocytes.
Catalog No. 50-256-1569