Learn More
Medchemexpress LLC BCI 100mg | 1245792-51-1 | 317.43 g·mol⁻¹ | C22H23NO | 100 MG
SDP
Supplier:  Medchemexpress LLC HY115502100MG
Encompass_Preferred
BCI is a small-molecule allosteric inhibitor of dual specificity phosphatases DUSP6 and DUSP1 supplied as a powder for laboratory research use. It has molecular formula C22H23NO, molecular weight 317.43 g·mol⁻¹, and is provided in solid pack sizes suitable for cell signaling and inflammatory disease research. Recommended storage guidance is provided for both powder and solvent solutions.
  • Allosteric inhibitor of DUSP6 and DUSP1 for research use.
  • High reported purity (about 99.7%).
  • Supplied as a powder solid for laboratory handling.
  • Available in multiple pack sizes, including 100 mg.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 2 years).
Catalog No. 50-003-07937