Learn More
Medchemexpress LLC HY-115502 10mg Medchemexpress, BCI CAS:1245792-51-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11550210MG
Encompass_Preferred
Medchemexpress, HY-115502 10mg BCI CAS:1245792-51-1 BCI is an allosteric inhibitor of dual specificity phosphatase (DUSP). BCI specifically inhibits DUSP6 and DUSP1 with EC50s of 13.3 and 8.0 μM in cells, respectively. BCI does not inhibit DUSP5. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9387