Learn More
Medchemexpress LLC BCI 25mg | 1245792-51-1 | 317.43 g·mol⁻¹ | C22H23NO | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11550225MG
Encompass_Preferred
BCI ((E)-BCI) is a small-molecule inhibitor of DUSP6 used for inflammatory disease and reactive oxygen species (ROS) research. It is provided as a high-purity solid suitable for in vitro studies and biochemical assays.
  • DUSP6 inhibitor active in cellular assays.
  • Demonstrates anti-inflammatory activity and reduction of ROS.
  • High purity (99.73% as supplied by manufacturer).
  • Molecular formula C22H23NO; molecular weight 317.43 g·mol⁻¹.
  • Available in small research quantities, including 25 mg vials.
Catalog No. 50-002-28027