Learn More
Selleck Chemical LLC BCIP E2924-100MG
SDP
Supplier:  Selleck Chemical LLC E2924100MG
Encompass_Preferred
BCIP (BCIP p-toluidine salt X-phosphate p-toluidine salt) is a synthetic chromogenic substrate used for the sensitive colorimetric detection of alkaline phosphatase activity
Catalog No. 50-000-40513