Learn More
Mabtech Inc BCIP- NBT SUBSTRATE 125ML
Supplier:  Mabtech Inc 365010
Encompass
This high quality precipitating BCIP/NBT-plus substrate is suitable for ELISpot assays with ALP conjugates.
Catalog No. NC0291099