Learn More
Research Products International Corp [X-Phos] [BCIP] 5-Bromo-4-chloro-3-indolyl-phosphate, disodium salt, 100 Milligrams
SDP
Supplier:  Research Products International Corp B740000.1
Encompass_Preferred
Colorimetric substrate for Alkaline phosphatase activity in blotting immunohistochemical and cytochemistry techniques. Forms a purple insoluble precipitate when used in conjunction with NBT.
Catalog No. 50-213-196