Learn More
Cayman Chemical BCIPp-toluIdIn salt 100mg
SDP
Supplier:  Cayman Chemical 34423100 MG
Encompass_Preferred
A colorimetric substrate for alkaline phosphatase; can be used to detect alkaline phosphatase activity by IHC or WB
Catalog No. 50-295-0975