Learn More
New England Biolabs, Inc. BciVI – 200 units
SDP
Supplier:  New England Biolabs, Inc. R0596S
Encompass
A restriction endonuclease that recognizes the sequence GTATCCNNNNN_N^.
Catalog No. 50-812-314