Learn More
TARGETMOL CHEMICALS INC BCL-B INHIBITOR 1 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T775715MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Bcl-B inhibitor 1 is a Bcl-B inhibitor with antitumor activity that binds to and inactivates pro-apoptotic proteins in the BH3 structural domain. purity: 97%
Catalog No. 50-257-9132