Learn More
Biotium BCL2-like 2(CPTC-BCL2L2-2), CF640R conjugate, 0.1mg/mL
Supplier:  Biotium BNC402265100
Encompass_Preferred
Bcl-2-like protein 2 (BCL2L2) is a member of the Bcl-2 protein family. Bcl-2-like protein 2 promotes cell survival and blocks dexamethasone-induced apoptosis. It also mediates survival of postmitotic Sertoli cells by suppressing death-promoting activity of BAX. Primary antibodies are available purified, or with a selection of fluorescent CF Dyes and other labels. CF Dyes offer exceptional brightness and photostability. Note: Conjugates of blue fluorescent dyes like CF405S and CF405M are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.
Catalog No. 50-006-72164