Learn More
Medchemexpress LLC BCL6-IN-5 | 2253878-09-8 | 99.8% | 396.27 | 25 MG

Supplier: Medchemexpress LLC 25MGHY136774
BCL6-IN-5 is a potent BCL6 inhibitor. It was extracted from patent WO2018215801A1, example 1n, and exhibits a pIC50 of 5.82. This product is intended for research use only.
- Potent BCL6 inhibitor
- Derived from patent WO2018215801A1
- Exhibits a pIC50 of 5.82
- Suitable for research applications
- High purity (99.82%)
- Available as a solid, light yellow to yellow in color
- Soluble in DMSO for in vitro studies
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.