Learn More
Medchemexpress LLC BCN-endo-PEG2-maleimide | 2675364-08-4 | 98.1% | 475.53 | C24H33N3O7 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY15631125MG
Encompass_Preferred
A bifunctional linker reagent containing an endo-bicyclononyne reactive group and a maleimide separated by a PEG2 spacer, designed for strain-promoted azide-alkyne cycloaddition and thiol conjugation in bioconjugation and drug-delivery workflows.
  • Contains endo-bicyclononyne and maleimide reactive groups.
  • Peg2 spacer provides flexibility and solubility.
  • Enables catalyst-free strain-promoted azide-alkyne cycloaddition (SPAAC) with azides.
  • Compatible with maleimide-mediated thiol conjugation.
  • High purity and defined molecular weight for reproducible conjugation.
Catalog No. 50-002-04335