Learn More
Medchemexpress LLC BCN-endo-PEG7-maleim |5mg
SDP
Supplier:  Medchemexpress LLC HY1563175MG
Encompass_Preferred
BCN-endo-PEG7-maleimide is an ADC Linker containing 7 PEG units BCN-endo-PEG7-maleimide contains the lyophilic bidentate macrocyclic ligand endo-BCN which can further synthesize macrocyclic complexes In click chemistry endo-BCN can react with molecules containing azide groups to form stable triazoles in the absence of catalysts
Catalog No. 50-002-04316