Learn More
Medchemexpress LLC Bcn-ss-nhs | 95.0% | 454.56 g/mol | C20H26N2O6S2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY13597325MG
Encompass_Preferred
BCN-SS-NHS is a cleavable antibody-drug conjugate (ADC) linker and click-chemistry reagent containing a bicyclo[6.1.0]nonyne (BCN) group and an N-hydroxysuccinimide (NHS) ester for conjugation. It is used for bioconjugation and ADC synthesis, enabling copper-free strain-promoted alkyne-azide cycloaddition (SPAAC) and disulfide-based release in reducing environments.
  • Cleavable disulfide linker for intracellular release
  • N-hydroxysuccinimide ester for efficient amine coupling
  • BCN moiety enables copper-free click chemistry (SPAAC)
  • High purity suitable for conjugation (95.0% by NMR)
  • Molecular weight 454.56 g/mol; formula C20H26N2O6S2
Catalog No. 50-002-02400