Learn More
Abcam BD 1008 dihydrobromide, delta1-receptor antagonist, 10MG
Supplier:  Abcam AB14455910MG
Encompass_Preferred
MW 463.1 Da, Purity >96%. Potent and selective δ1-receptor antagonist (Ki = 0.34 nM). Therapeutic agent. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5631