Learn More
Abcam BD 1063 dihydrochloride, sigma-1 antagonist, 10MG
Supplier:  Abcam AB14132310MG
Encompass_Preferred
MW 346.1 Da, Purity >99%. Potent selective σ1 receptor antagonist (Ki values are 9 and 449 nM at σ1 and σ2 receptors, respectively). Shown to prevent hyperlocomotion, active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6679