Learn More
Medchemexpress LLC BD-4 Human 100ug
SDP
Supplier:  Medchemexpress LLC HYP7140100UG
Encompass_Preferred
BD-4 Protein, Human is a recombinant human antimicrobial peptide provided for research applications. It displays an inducible, salt-sensitive spectrum of antimicrobial activity and is intended for in vitro studies of host defense peptides, innate immunity, and antimicrobial mechanisms. The material is supplied at research-scale quantity with reported high purity and is shipped refrigerated to preserve activity.
  • Recombinant human antimicrobial peptide suitable for in vitro research.
  • Inducible, salt-sensitive antimicrobial activity useful for host defense studies.
  • High reported purity supports biochemical and functional assays.
  • Reported molecular weight approximately 5-11 kDa for experimental planning.
  • Supplied in research-scale quantities and shipped refrigerated to maintain integrity.
Catalog No. 50-001-95651