Learn More
BD Cell Analysis 3P 7-Color Setup Beads, use on a BD FACSCanto flow cytometer, 25 tests
Supplier:  BD Cell Analysis 3P 335775
Encompass
BD FACS 7-color setup beads are for in vitro diagnostic use on a BD FACSCanto flow cytometer with BD FACSCanto software. The beads are used to adjust detector voltages, to set fluorescence compensation, and to monitor daily instrument performance. Item includes 25 tests.
Catalog No. 50-620-008