Learn More
BD Cell Analysis 3P BD FACSFlow Sheath Fluid, 20 liter
Supplier: BD Cell Analysis 3P 342003
BD FACSFlow sheath fluid - 20L. Physical and chemical properties: Liquid form, clear in color with a pH value between 7.8-8.2. Boiling point/range: 100C (212F). Density at 20C is 1.0145g/cm3