Learn More
BD Cell Analysis 3P Retic-COUNT™ reagent 200 tests
Supplier:  BD Cell Analysis 3P 349204
Encompass
Sufficient for 200 tests, BD Retic-Count reagent is provided in a 200-mL bottle. The solution contains Thiazole Orange in phosphate-buffered saline (PBS) with 0.1% sodium azide.
Catalog No. 50-620-716