Learn More
Takara Bio BD TALON XTRACTOR BUFFER 500
SDP
Supplier:  Takara Bio 635625
Encompass
BD Talon xTractor Buffer,500mL,Ready-To-Use Format, Mild, Non-Denaturing Conditions, Optimized for Purifying Poly-His Tag Proteins, High Efficiency Extraction, Preparation and Storage: -20 deg.C (DNase I), 4 deg.C for All Other Components
Catalog No. NC9090696