Learn More
Strem, An Ascensus Company CAS# [27804-64-4]. 25g. Bis(diethylamino)silane, 99% (99.999%-Si) BDEAS PURATREM.
SDP
Supplier:  Strem, An Ascensus Company 98881025G
Encompass
CAS# [27804-64-4]. 25g. Bis(diethylamino)silane, 99% (99.999%-Si) BDEAS PURATREM. . Molecular Weight: 174.36. Molecular Formula: SiH2[N(CH2CH3)2]2. Color/form: colorless liq. Strem# 98-8810. www.strem.com/catalog/v3/98-8810/
Catalog No. 50-197-9374