Learn More
Sage Science Inc Reagent Replacement Kit, BluePippin™ Gel Cassettes
SDP
Supplier:  Sage Science Inc RBF2010
Encompass
DNA fragments (Fluorescein label) with Ficoll in water in tube:<200ug DNA with 30% Ficoll in Tris-EDTA buffer (~500 ul vol.)Tris-TAPS Buffer (~30 ml in 60ml bottle)
Catalog No. NC1143349