Learn More
Andwin Scientific BDH Buffer PH 4, Red, 500mL
SDP
Supplier:  Andwin Scientific BDH5018500ML
Encompass_Preferred
The BDH pH reference standard buffer is a clear, light red, odorless, liquid solution. This contains hydrochloric acid, potassium chloride, and deionized water. It is classified to have a 3.99-4.01 pH at 25.0 C. This product is packaged and sold in a 500 mL poly bottle.
Catalog No. NC0491791