Learn More
Electron Microscopy Sciences BDMA, Benzyldimethylamine 100 ML

Supplier: Electron Microscopy Sciences 11400
(N-Benzyl-N, N-Dimethylamine)
CAS #103-83-3 C₆H₅CH₂N(CH₃)₂ F.W. 135.21 g/mol
Boiling point: 177-180 °C
N-Benzyl-N, N-Dimethylamine is an amine accelerator for polymerization of epoxy resins. A direct alternative to DMP-30 with a longer shelf life and better penetration.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.