Learn More
Medchemexpress LLC BDNF Human CHO 10ug
SDP
Supplier:  Medchemexpress LLC HYP711610UG
Encompass_Preferred
Recombinant human brain-derived neurotrophic factor (BDNF), expressed in CHO cells and supplied lyophilized for research use. The material corresponds to residues H129-R247, is >95% pure by reducing SDS-PAGE, and is formulated from 0.22 μm filtered PBS. It is intended for in vitro studies of neuronal survival, differentiation, and receptor signaling; store lyophilized at -20°C and aliquot for long-term storage at -80°C.
  • Expressed in CHO cells for mammalian post-translational modifications.
  • Covers residues H129-R247 relevant to receptor binding.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized from a 0.22 μm filtered PBS solution for stability.
  • Suitable for in vitro studies of neuronal survival, differentiation, and signaling.
  • Stable when frozen; aliquot and store at -20°C or -80°C for extended storage.
Catalog No. 50-001-97234