Learn More
Medchemexpress LLC BDPI 250mg
SDP
Supplier:  Medchemexpress LLC 250MGHYW112239
Encompass_Preferred
BDPI is the a BODIPY derivative BDPI reveals a high O2 quantum yield and exhibits phototoxicity that inhibits HeLa with an IC50 of 0 6 g/mL BDPI nanoarticles can be taken up by HeLa producing fluorescence signals in the cells and can be used as cell imaging agent BDPI nanoarticles exhibit antitumor efficacy in mouse models[1]
Catalog No. 50-002-05775