Learn More
Abcam BDSF (cis-2-dodecenoic acid), Diffusible fatty acid signal molecule, 10MG
Supplier:  Abcam AB14534110MG
Encompass_Preferred
MW 198.3 Da, Purity >95%. Diffusible fatty acid signal molecule. Acts as a bacterial virulence factor. Activates DSF-dependent responses. Inhibits germ tube formation in Candida albicans. Shows antifungal effects in vivo..
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5282