Learn More
BD Cell Analysis 3P Accudrop Beads, single population of 6-µm particles, 25 tests
Supplier:  BD Cell Analysis 3P 345249
Encompass
Accudrop Beads for flow cytometry; BD FACS brand; 25 tests. This product contains a single population of 6-um particles. Every particle contains a fluorophore that is excited at 670 nm and emits at 750 nm.
Catalog No. 50-620-551