Learn More
Chemglass Life Sciences Beaker, 2L, 316L SS, 6.3" H x 5.51" ID
SDP
Supplier:  Chemglass Life Sciences CG1109X2000
Encompass
Beakers are made from 316L stainless steel and available in capacities of 1, 2, 3, and 5 liters. Beakers have solid bar sanitary handle, easy-pouring beaker-spout, and graduation marks. All beakers have a seamless sanitary interior and standard 10 Ra finish. Features: Beakers have solid bar sanitary handle. Easy-pouring beaker-spout. Graduation Subdivision lines/marks (without numbers). 316L Polished 10 Ra Finish.
Catalog No. 50-009-80674