Learn More
SAINT GOBAIN PERF PLASTICS POESTENKIL Chemware&trade, PTFE Griffin Beakers, Capacity, 250mL
Supplier:  SAINT GOBAIN PERF PLASTICS POESTENKIL D1069038
Encompass_Preferred
Noncontaminating and inert to highly corrosive reagents such as hydrofluoric acid. Nonwettable, unbreakable and heat resistantEasy to cleanBroad spout for easy pouringCan be heated on hotplate up to 288°C (550°F) when proper care is exercisedSpecially molded bottom allows rapid heat transferUnlike most PTFE beakers, these will boil water when coveredApplications:Ideal for digestion
Catalog No. 50-206-7750