Learn More
Wilmad Labglass BEAKER JACKETED 400ML
Supplier:  Wilmad Labglass LG3300104
Encompass
Beaker, Jacketed. Capacity: 400mL. Fully jacketed to all temperature control of contents. Similar to a Griffin beaker, except without a pouring spout.
Catalog No. 50-806-377
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.