Learn More
Chemglass Life Sciences Chemglass 30 mL Borosilicate Griffin Beaker, Low Form, Graduated
SDP
Supplier:  Chemglass Life Sciences CG804830
Encompass_Preferred
Chemglass low-form Griffin beakers are general-purpose laboratory beakers for mixing, heating, and approximate volume measurement. This 30 mL beaker is made from borosilicate glass and features a pouring spout and graduated markings for convenient, small-volume work.
  • 30 mL capacity suitable for small-volume measurements and heating.
  • Low-form Griffin style for easy stirring and handling.
  • Borosilicate glass construction for thermal shock resistance.
  • Graduated scale for approximate volume measurement.
  • Pouring spout for controlled transfer.
  • Uniform wall thickness for strength and durability.
  • Available in pack quantities for laboratory stocking.
Catalog No. NC2943926