Learn More
Chemglass Life Sciences Beaker, Griffin, Polypropylene, 5 Liter, Graduated
SDP
Supplier:  Chemglass Life Sciences CG1108L01
Encompass_Preferred
Beaker, Griffin, Polypropylene, 5 Liter, Graduated. Graduated polypropylene beakers with flared tops and a pouring spouts. Steam autoclavable at 121°C.
Catalog No. 50-872-720