Learn More
Chemglass Life Sciences Beaker, PTFE, 500mL, 125mm OAH X 80mm OD
SDP
Supplier:  Chemglass Life Sciences CG110904
Encompass_Preferred
Beaker, PTFE, 500mL, 120mm OAH X 80mm OD. Chemically inert PTFE beakers can withstand temperatures up to 260¿C.
Catalog No. 50-872-725