Learn More
Ace Glass, Inc. PTFE BEAKER, 600ML
SDP
Supplier:  Ace Glass, Inc. 550011
Encompass
600mL flat-bottom beaker made from molded virgin PTFE, with pour spout. Inert, with smooth internal finish.
Catalog No. 50-971-887