Learn More
Chemglass Life Sciences Beaker, Quartz, 600mL, 119mm OAH X 90mm OD
SDP
Supplier:  Chemglass Life Sciences CG111005
Encompass_Preferred
Beaker, Quartz, 600mL, 119mm OAH X 90mm OD. Low form beaker made of quartz. Non-graduated with pouring spout. Beaker can be used at temperatures to 1000°C.
Catalog No. 50-871-937