Learn More
Bellco Glass, Inc. Manufactured from Type 1 Class A Borosilicate Glass with a low coefficient of expansion. ideal for general laboratory use. Features an easy pour spout, white permanent graduations and large marking square. Size: 600ml Dimensions: 88x123mm
SDP
Supplier:  Bellco Glass, Inc. 562100600
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass with a low coefficient of expansion. ideal for general laboratory use. Features an easy pour spout, white permanent graduations and large marking square. Size: 600ml Dimensions: 88x123mm
Catalog No. 50-194-5698