Learn More
Chemglass Life Sciences Deflecting Beam Torque Wrench, MAX 60 in-lbs
SDP
Supplier:  Chemglass Life Sciences CG1963B10
Encompass_Preferred
Deflecting Beam Torque Wrench, MAX 60 in-lbs. Deflecting beam torque wrench. Directly measures torque when tightening fasteners. Pointer moves clockwise or counter clockwise to measure torque. Comfortable, palm fit handle provides a natural grip for smooth accurate pull. Wrench has 1/4" drive and the overall length is 11". PLEASE NOTE: FOR USE WITH CG-1968 BEADED PIPE COUPLINGS.
Catalog No. CG1963B10