Learn More
Wilmad Labglass BEARING CHSPKE 24/40
Supplier:  Wilmad Labglass LG9470200
Encompass_Preferred
Stirrer Bearing, Chesapeake, 10mm. Joint Size: [ST] 24/40. Grease-free PTFE Stirrer Bearing designed for moderate vacuum applications or for use at atmospheric pressure.
Catalog No. 50-806-280