Learn More
Medchemexpress LLC Bebt-109 | 2050906-40-4 | 98.6% | 516.59 g·mol⁻¹ | C27H32N8O3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW423595100MG
Encompass_Preferred
BEBT-109 is a research-grade, pan-mutant-selective epidermal growth factor receptor (EGFR) inhibitor designed for preclinical studies of mutant EGFR-driven non-small cell lung cancer. It is supplied as a white to off-white solid with a molecular formula of C27H32N8O3, molecular weight 516.59 g·mol⁻¹, and a reported purity of approximately 98.6%.
  • Pan-mutant-selective EGFR inhibitor for mutant EGFR-driven NSCLC research.
  • High reported purity suitable for biochemical and cellular assays.
  • Supplied as a stable solid for convenient storage and handling.
  • Documented storage conditions support long-term sample integrity.
  • Characterized molecular properties aid reproducible experimental design.
Catalog No. 50-002-81327