Learn More
Medchemexpress LLC Beclomethasone 17-propionate | 5534-18-9 | 99.5% | 464.98 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY13623910MM/1ML
Encompass_Preferred
Beclomethasone 17-propionate is an active metabolite of Beclomethasone dipropionate and acts as a glucocorticoid receptor (GR) agonist. It demonstrates a higher affinity for GR compared to Beclomethasone dipropionate and is effective in suppressing cytokine production in lung macrophages of individuals with chronic obstructive pulmonary disease (COPD).
  • Metabolism of beclomethasone dipropionate to 17-BMP is a crucial activation step.
  • Inhibits LPS-stimulated CXCL8, TNFα, and IL-6.
  • Upregulates GR-dependent genes FKBP51 and GILZ.
Catalog No. 50-003-50952