Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beclomethasone 17-propionate-d5 | >95.0% | 470.01 | C25H28D5ClO6 | 1 MG
SDP

Supplier:  Medchemexpress LLC HYW6540661MG

Encompass_Preferred

Beclomethasone 17-Propionate-d5 is the deuterium-labeled (d5) analogue of beclomethasone 17-propionate supplied for research use as a stable isotope internal standard and tracer in analytical and drug-development workflows.

  • Deuterium-labeled internal standard for quantitative LC-MS assays.
  • Molecular formula C25H28D5ClO6; molecular weight 470.01.
  • Purity >95% by HPLC.
  • Intended for research use only; follow Certificate of Analysis for storage and handling.

Catalog No. 50-002-84840


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.