Learn More
Selleck Chemical LLC Beclomethasone dipropionate S3078-5mg
SDP
Supplier:  Selleck Chemical LLC S30785MG
Encompass_Preferred
Beclomethasone dipropionate(Beclovent Beconase Vancenase) is a potent glucocorticoid steroid used for the treatment of rhinitis and sinusitis
Catalog No. 50-311-1327