Learn More
Apexbio Technology LLC Beclomethasone dipropionate 5534-09-8 50mg
SDP
Supplier:  Apexbio Technology LLC B124850
Encompass_Preferred
Beclomethasone dipropionate (CAS 5534-09-8) is a small-molecule agonist targeting glucocorticoid receptors It is designed to modulate glucocorticoid receptor activity thereby suppressing inflammatory responses Beclomethasone dipropionate exerts its biological activity primarily through binding to glucocorticoid receptors leading to modulation of gene expression and inhibition of inflammatory mediator release Based on these pharmacological properties beclomethasone dipropionate holds research potential in the study of respiratory conditions such as asthma and allergic rhinitis as well as in evaluating glucocorticoid receptor interactions and therapeutic response in preclinical and clinical models
Catalog No. 50-000-92399